4-(5,5-Dimethyl-1,3-dioxan-2-yl)aniline ≥98%

Supplier: Ambeed
Warning

A1227387-100MG A1227387-1G
77866-866EA 51 USD
77866-866 77866-867
4-(5,5-Dimethyl-1,3-dioxan-2-yl)aniline ≥98%
4-(5,5-Dimethyl-1,3-dioxan-2-yl)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂
CAS Number: 107708-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR