11-Oxo-N-((2-(trifluoromethyl)thiazol-5-yl)methyl)-10,11-dihydrodibenzo[b,f][1,4]thiazepine-8-carboxamide 5,5-dioxide ≥97%

Supplier: Ambeed

A1227262-10MG A1227262-1MG A1227262-5MG A1227262-100MG A1227262-25MG A1227262-50MG
77596-456EA 138.38 USD
77596-456 77596-458 77596-460 77866-862 77866-863 77866-864
11-Oxo-N-((2-(trifluoromethyl)thiazol-5-yl)methyl)-10,11-dihydrodibenzo[b,f][1,4]thiazepine-8-carboxamide 5,5-dioxide ≥97%
11-Oxo-N-((2-(trifluoromethyl)thiazol-5-yl)methyl)-10,11-dihydrodibenzo[b,f][1,4]thiazepine-8-carboxamide 5,5-dioxide

Precaución: for Research Use Only

Formula: C₁₉H₁₂F₃N₃O₄S₂
MDL Number: MFCD32856555
CAS Number: 2090064-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR