2-(2-(Prop-2-yn-1-yloxy)ethoxy)ethyl 4-methylbenzenesulfonate ≥97%
Supplier: Ambeed
|
Danger
|
Synonyms:
Tos-PEG2-O-Propargyl
A1226055-100MG
A1226055-1G
A1226055-250MG
A1226055-5G
77596-444EA
37.93
USD
77596-444
77596-446
77596-448
77621-136
2-(2-(Prop-2-yn-1-yloxy)ethoxy)ethyl 4-methylbenzenesulfonate ≥97%
2-(2-(Prop-2-yn-1-yloxy)ethoxy)ethyl 4-methylbenzenesulfonate
Precaución: for Research Use Only
Formula:
C₁₄H₁₈O₅S |
MDL Number:
MFCD20134051 CAS Number: 1119249-30-7 UN: 2810 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...