2,2-Dimethyl-4-oxo-3,8,11,14,17,20-hexaoxa-5-azadocosan-22-oic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: t-Boc-N-amido-PEG5-acetic acid

A1226005-100MG A1226005-1G A1226005-5G A1226005-250MG
77596-438EA 45.1 USD
77596-438 77596-440 77596-442 77621-134
2,2-Dimethyl-4-oxo-3,8,11,14,17,20-hexaoxa-5-azadocosan-22-oic acid ≥98%
2,2-Dimethyl-4-oxo-3,8,11,14,17,20-hexaoxa-5-azadocosan-22-oic acid

Precaución: for Research Use Only

Formula: C₁₇H₃₃NO₉
MDL Number: MFCD31656900
CAS Number: 890152-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR