endo-BCN-PEG3-acid ≥98%

Supplier: Ambeed
Warning

A1225964-250MG A1225964-50MG A1225964-100MG
77571-374EA 792 USD
77571-374 77866-840 77866-839
endo-BCN-PEG3-acid ≥98%
endo-BCN-PEG3-acid

Precaución: for Research Use Only

Formula: C₂₀H₃₁NO₇
MDL Number: MFCD31692198
CAS Number: 1807501-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR