(2S,4S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-methylpyrrolidine-2-carboxylic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-cis-4-Me-Pro-OH

A1224552-1G A1224552-100MG A1224552-250MG
77571-364EA 391 USD
77571-364 77866-823 77866-824
(2S,4S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-methylpyrrolidine-2-carboxylic acid ≥95%
(2S,4S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-methylpyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
MDL Number: MFCD31716148
CAS Number: 1228577-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR