3-(6-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione ≥95%

Supplier: Ambeed

A1223862-250MG A1223862-100MG
77596-434EA 145.55 USD
77596-434 77866-814
3-(6-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione ≥95%
3-(6-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione

Precaución: for Research Use Only

Formula: C₁₃H₁₃N₃O₃
MDL Number: MFCD31692699
CAS Number: 191732-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR