5-Isopropylpyridin-3-amine ≥95%

Supplier: Ambeed
Warning

A122271-100MG A122271-250MG
77889-842EA 52 USD
77889-842 77889-843
5-Isopropylpyridin-3-amine ≥95%
5-Isopropylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
MDL Number: MFCD18256499
CAS Number: 1256821-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR