(S)-3-Amino-4-methylpentanoic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: H-β-HoVal-OH

A122211-10G A122211-1G
77614-960EA 157.59 USD
77614-960 77745-895
(S)-3-Amino-4-methylpentanoic acid ≥97%
(S)-3-Amino-4-methylpentanoic acid

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
MDL Number: MFCD01076233
CAS Number: 40469-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR