(4,4-Difluorocyclohexyl)hydrazine dihydrochloride ≥98%

Supplier: Ambeed

A1220872-1G A1220872-100MG A1220872-250MG
77571-346EA 220 USD
77571-346 77866-803 77866-804
(4,4-Difluorocyclohexyl)hydrazine dihydrochloride ≥98%
(4,4-Difluorocyclohexyl)hydrazine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄Cl₂F₂N₂
MW: 223.09 g/mol
MDL Number: MFCD30145539
CAS Number: 1911568-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR