Methyl 7-bromothieno[2,3-c]pyridine-2-carboxylate ≥95%

Supplier: Ambeed
Warning

A122084-100MG A122084-1G A122084-250MG
77614-952EA 135.96 USD
77614-952 77614-954 77614-956
Methyl 7-bromothieno[2,3-c]pyridine-2-carboxylate ≥95%
Methyl 7-bromothieno[2,3-c]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂S
MW: 272.1200 g/mol
CAS Number: 2092325-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR