(4S,4'S,5R,5'R)-2,2'-(Cyclohexane-1,1-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) ≥97%, ee 99%

Supplier: Ambeed
Warning

A1220168-1G A1220168-250MG A1220168-100MG
77621-084EA 384.19 USD
77621-084 77621-086 77866-789
(4S,4'S,5R,5'R)-2,2'-(Cyclohexane-1,1-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) ≥97%, ee 99%
(4S,4'S,5R,5'R)-2,2'-(Cyclohexane-1,1-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₃₆H₃₄N₂O₂
MW: 526.660 g/mol
MDL Number: MFCD32201210
CAS Number: 2634687-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR