5,6-Dihydrouridine ≥97%

Supplier: Ambeed
Warning

A1220137-1MG A1220137-10MG A1220137-5MG A1220137-100MG A1220137-25MG A1220137-50MG
77596-420EA 43.05 USD
77596-420 77621-070 77621-072 77866-784 77866-785 77866-786
5,6-Dihydrouridine ≥97%
5,6-Dihydrouridine

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₆
MDL Number: MFCD00047490
CAS Number: 5627-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR