2,6-Bis((diphenylphosphanyl)methyl)pyridine ≥97%

Supplier: Ambeed
Warning

A1220051-250MG A1220051-1G
77571-332EA 113.3 USD
77571-332 77621-064
2,6-Bis((diphenylphosphanyl)methyl)pyridine ≥97%
2,6-Bis((diphenylphosphanyl)methyl)pyridine

Precaución: for Research Use Only

Formula: C₃₁H₂₇NP₂
MW: 475.5000 g/mol
MDL Number: MFCD13178323
CAS Number: 73892-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR