(11bR)-N,N-Bis((S)-1-phenylethyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine ≥97%, ee 99%

Supplier: Ambeed
Warning

A1220017-100MG A1220017-250MG
77592-378EA 43.05 USD
77592-378 77621-060
(11bR)-N,N-Bis((S)-1-phenylethyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine ≥97%, ee 99%
(11bR)-N,N-Bis((S)-1-phenylethyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine

Precaución: for Research Use Only

Formula: C₃₆H₃₈NO₂P
MW: 547.67 g/mol
CAS Number: 1021439-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR