(S)-6,6'-Diiodo-2,2',3,3'-tetrahydro-1,1'-spirobi[indene]-7,7'-diol ≥97%

Supplier: Ambeed
Warning

A1219953-1G A1219953-250MG
77621-050EA 899.19 USD
77621-050 77621-052
(S)-6,6'-Diiodo-2,2',3,3'-tetrahydro-1,1'-spirobi[indene]-7,7'-diol ≥97%
(S)-6,6'-Diiodo-2,2',3,3'-tetrahydro-1,1'-spirobi[indene]-7,7'-diol

Precaución: for Research Use Only

Formula: C₁₇H₁₄I₂O₂
MW: 504.1 g/mol
CAS Number: 1258326-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR