(R)-6,6'-Diiodo-2,2',3,3'-tetrahydro-1,1'-spirobi[indene]-7,7'-diol ≥97%

Supplier: Ambeed
Warning

A1219952-100MG A1219952-1G A1219952-250MG A1219952-5G
77621-044EA 185.4 USD
77621-044 77621-046 77621-048 77866-764
(R)-6,6'-Diiodo-2,2',3,3'-tetrahydro-1,1'-spirobi[indene]-7,7'-diol ≥97%
(R)-6,6'-Diiodo-2,2',3,3'-tetrahydro-1,1'-spirobi[indene]-7,7'-diol

Precaución: for Research Use Only

Formula: C₁₇H₁₄I₂O₂
MW: 504.1 g/mol
CAS Number: 1435948-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR