(4S,4'S)-2,2'-(Cyclobutane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) ≥97%

Supplier: Ambeed
Warning

A1219918-100MG A1219918-250MG
77621-026EA 145.23 USD
77621-026 77621-028
(4S,4'S)-2,2'-(Cyclobutane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) ≥97%
(4S,4'S)-2,2'-(Cyclobutane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₁₈H₃₀N₂O₂
MW: 306.4430 g/mol
CAS Number: 298693-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR