3-Acetoxy-1-propenylboronic acid pinacol ester ≥97%

Supplier: Ambeed
Warning

A1219778-250MG A1219778-1G A1219778-5G
77571-314EA 63 USD
77571-314 77866-743 77866-744
3-Acetoxy-1-propenylboronic acid pinacol ester ≥97%
3-Acetoxy-1-propenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₉BO₄
MDL Number: MFCD03788751
CAS Number: 161395-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR