(E)-3-(2-Cyano-phenyl)-acrylic acid ≥98%

Supplier: Ambeed
Warning

A1219517-250MG A1219517-1G
77621-002EA 38.11 USD
77621-002 77866-735
(E)-3-(2-Cyano-phenyl)-acrylic acid ≥98%
(E)-3-(2-Cyano-phenyl)-acrylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MDL Number: MFCD00189382
CAS Number: 2886-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR