Cyclooct-4-en-1-yl (3-aminopropyl)carbamate hydrochloride ≥97%

Supplier: Ambeed
Warning

A1219442-1G A1219442-250MG A1219442-50MG A1219442-100MG
77620-996EA 683.92 USD
77620-996 77620-998 77621-000 77620-994
Cyclooct-4-en-1-yl (3-aminopropyl)carbamate hydrochloride ≥97%
Cyclooct-4-en-1-yl (3-aminopropyl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₂₃ClN₂O₂
MW: 262.7762 g/mol
MDL Number: MFCD32665932
CAS Number: 1609659-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR