Sodium 4,4-difluorocyclohexane-1-sulfinate ≥98%

Supplier: Ambeed
Warning

A1218056-1G A1218056-250MG
77592-374EA 552.48 USD
77592-374 77620-976
Sodium 4,4-difluorocyclohexane-1-sulfinate ≥98%
Sodium 4,4-difluorocyclohexane-1-sulfinate

Precaución: for Research Use Only

Formula: C₆H₉F₂NaO₂S
MW: 206.19 g/mol
MDL Number: MFCD28124334
CAS Number: 1622013-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR