(4-Ethynylphenyl)diphenylphosphane ≥97%

Supplier: Ambeed
Danger

A1217974-1G A1217974-250MG A1217974-100MG
77596-394EA 699.05 USD
77596-394 77596-396 77620-972
(4-Ethynylphenyl)diphenylphosphane ≥97%
(4-Ethynylphenyl)diphenylphosphane

Precaución: for Research Use Only

Formula: C₂₀H₁₅P
MW: 286.31 g/mol
CAS Number: 359017-78-0
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR