Methyl-5-nitro-1H-indazole-3-carboxylate ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A121715-1G A121715-250MG A121715-5G
77683-096EA 104 USD
77683-096 77889-835 77889-836
Methyl-5-nitro-1H-indazole-3-carboxylate ≥95%, Ambeed.Inc
Methyl 5-nitro-1H-indazole-3-carboxylate

Precaución: For research use only.

Formula: C₉H₇N₃O₄
MDL Number: MFCD07371594
CAS Number: 78155-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR