(R)-4-(3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)-N-(3-chloropyridin-2-yl)-N-(piperidin-3-yl)benzamide xhydrochloride ≥98%

Supplier: Ambeed

Synonyms: PF-06446846 HCl

A1216950-100MG A1216950-250MG A1216950-50MG
77866-722EA 504 USD
77866-722 77866-723 77866-724
(R)-4-(3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)-N-(3-chloropyridin-2-yl)-N-(piperidin-3-yl)benzamide xhydrochloride ≥98%
(R)-4-(3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)-N-(3-chloropyridin-2-yl)-N-(piperidin-3-yl)benzamide xhydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₁Cl₂N₇O
MW: 470.35 g/mol
MDL Number: MFCD31693841
CAS Number: 1632250-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR