2-(4-((4R,5S)-4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydro-1H-imidazole-1-carbonyl)-2-oxopiperazin-1-yl)acetic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: (4R,5S)-Nutlin carboxylic acid

A1216861-50MG A1216861-100MG
77596-392EA 379.25 USD
77596-392 77620-970
2-(4-((4R,5S)-4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydro-1H-imidazole-1-carbonyl)-2-oxopiperazin-1-yl)acetic acid ≥95%
2-(4-((4R,5S)-4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydro-1H-imidazole-1-carbonyl)-2-oxopiperazin-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₃₂H₃₂Cl₂N₄O₆
MW: 639.53 g/mol
MDL Number: MFCD32197284
CAS Number: 2306390-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR