N-(3-(6-Amino-5-(2-(N-methylacrylamido)ethoxy)pyrimidin-4-yl)-5-fluoro-2-methylphenyl)-4-cyclopropyl-2-fluorobenzamide ≥98%

Supplier: Ambeed
Danger

Synonyms: Remibrutinib

A1216695-100MG A1216695-25MG A1216695-50MG A1216695-250MG
77620-958EA 572.68 USD
77620-958 77620-960 77620-962 77866-713
N-(3-(6-Amino-5-(2-(N-methylacrylamido)ethoxy)pyrimidin-4-yl)-5-fluoro-2-methylphenyl)-4-cyclopropyl-2-fluorobenzamide ≥98%
N-(3-(6-Amino-5-(2-(N-methylacrylamido)ethoxy)pyrimidin-4-yl)-5-fluoro-2-methylphenyl)-4-cyclopropyl-2-fluorobenzamide

Precaución: for Research Use Only

Formula: C₂₇H₂₇F₂N₅O₃
MDL Number: MFCD32215329
CAS Number: 1787294-07-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR