4-(1-(5-(4,5-Dimethyl-1H-pyrazol-3-yl)-2,4-dimethylbenzoyl)azetidin-3-yl)benzonitrile ≥98%

Supplier: Ambeed
Warning

A1216683-50MG A1216683-10MG A1216683-25MG A1216683-100MG
77620-956EA 332.94 USD
77620-956 77620-952 77620-954 77866-712
4-(1-(5-(4,5-Dimethyl-1H-pyrazol-3-yl)-2,4-dimethylbenzoyl)azetidin-3-yl)benzonitrile ≥98%
4-(1-(5-(4,5-Dimethyl-1H-pyrazol-3-yl)-2,4-dimethylbenzoyl)azetidin-3-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₂₄H₂₄N₄O
MDL Number: MFCD30534126
CAS Number: 1533438-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR