(R)-4-((4-(((4-((Tetrahydrofuran-3-yl)oxy)benzo[d]isoxazol-3-yl)oxy)methyl)piperidin-1-yl)methyl)tetrahydro-2H-pyran-4-ol ≥98%

Supplier: Ambeed

Synonyms: PF-04995274

A1216666-10MG A1216666-1MG A1216666-5MG
77596-356EA 127.1 USD
77596-356 77596-358 77596-360
(R)-4-((4-(((4-((Tetrahydrofuran-3-yl)oxy)benzo[d]isoxazol-3-yl)oxy)methyl)piperidin-1-yl)methyl)tetrahydro-2H-pyran-4-ol ≥98%
(R)-4-((4-(((4-((Tetrahydrofuran-3-yl)oxy)benzo[d]isoxazol-3-yl)oxy)methyl)piperidin-1-yl)methyl)tetrahydro-2H-pyran-4-ol

Precaución: for Research Use Only

Formula: C₂₃H₃₂N₂O₆
MW: 432.51 g/mol
MDL Number: MFCD29079009
CAS Number: 1331782-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR