(R)-7-Chloro-N-((4,6-dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-2-(trans-4-(dimethylamino)cyclohexyl)-2,4-dimethylbenzo[d][1,3]dioxole-5-carboxamide 4-methylbenzenesulfonate ≥98%

Supplier: Ambeed
Warning

A1216638-10MG A1216638-1MG A1216638-5MG A1216638-50MG A1216638-25MG
77620-942EA 287.37 USD
77620-942 77620-944 77620-946 77866-710 77866-709
(R)-7-Chloro-N-((4,6-dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-2-(trans-4-(dimethylamino)cyclohexyl)-2,4-dimethylbenzo[d][1,3]dioxole-5-carboxamide 4-methylbenzenesulfonate ≥98%
(R)-7-Chloro-N-((4,6-dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-2-(trans-4-(dimethylamino)cyclohexyl)-2,4-dimethylbenzo[d][1,3]dioxole-5-carboxamide 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₃₃H₄₂ClN₃O₇S
MDL Number: MFCD32197179
CAS Number: 1809336-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR