5-(Aminomethyl)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride ≥97%

Supplier: Ambeed

Synonyms: Thalidomide-5-CH2-NH2 HCl

A1215841-250MG A1215841-1G
77571-286EA 429 USD
77571-286 77866-707
5-(Aminomethyl)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride ≥97%
5-(Aminomethyl)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₄ClN₃O₄
MW: 323.73 g/mol
MDL Number: MFCD32666042
CAS Number: 1010100-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR