2-Hydroxy-3-nitro-5-picoline ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-3-nitro-2-pyridone, 2-Hydroxy-5-methyl-3-nitropyridine, 6-Hydroxy-5-nitro-3-picoline, 5-Methyl-3-nitro-2-pyridinol, 5-Methyl-3-nitro-2(1H)-pyridone

A121395-25G A121395-5G A121395-100G
77670-718EA 47 USD
77670-718 77670-720 77882-984
2-Hydroxy-3-nitro-5-picoline ≥98%
2-Hydroxy-3-nitro-5-picoline

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.13
MDL Number: MFCD02070021
CAS Number: 7464-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR