(R)-5-Methylpyrrolidin-2-one ≥98%

Supplier: Ambeed
Warning

A121341-250MG A121341-100MG A121341-1G A121341-5G
77614-944EA 127.72 USD
77614-944 77747-881 77747-882 77747-883
(R)-5-Methylpyrrolidin-2-one ≥98%
(R)-5-Methylpyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₅H₉NO
MW: 99.13 g/mol
MDL Number: MFCD19216883
CAS Number: 21395-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR