Iron(III) 2-methyl-4-oxo-4H-pyran-3-olate ≥97%

Supplier: Ambeed
Warning

Synonyms: Ferric maltol

A1213222-1G A1213222-250MG A1213222-5G
77620-900EA 72.1 USD
77620-900 77620-902 77620-904
Iron(III) 2-methyl-4-oxo-4H-pyran-3-olate ≥97%
Iron(III) 2-methyl-4-oxo-4H-pyran-3-olate

Precaución: for Research Use Only

Formula: C₁₈H₁₅FeO₉
MW: 431.1513 g/mol
MDL Number: MFCD33402141
CAS Number: 33725-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR