Methyl 11-aminoundecanoate hydrochloride ≥97%

Supplier: Ambeed
Warning

A1213218-1G A1213218-5G A1213218-250MG
77596-332EA 17.43 USD
77596-332 77596-334 77620-898
Methyl 11-aminoundecanoate hydrochloride ≥97%
Methyl 11-aminoundecanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₂₆ClNO₂
MW: 251.7933 g/mol
MDL Number: MFCD03161190
CAS Number: 29833-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR