1-Phenylsulfonyl-7-azaindole-4-boronic acid pinacol ester 98+%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 1-(Phenylsulphonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine

A121262-100MG A121262-1G A121262-5G A121262-250MG
77683-089EA 65 USD
77683-089 77683-090 77683-091 77891-865
1-Phenylsulfonyl-7-azaindole-4-boronic acid pinacol ester 98+%, Ambeed.Inc
1-Phenylsulfonyl-7-azaindole-4-boronic acid pinacol ester

Precaución: For research use only.

Formula: C₁₉H₂₁BN₂O₄S
MDL Number: MFCD10574997
CAS Number: 942919-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR