2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalonitrile ≥98%

Supplier: Ambeed
Warning

A1212412-1G A1212412-100MG A1212412-250MG A1212412-5G
77592-364EA 448.95 USD
77592-364 77620-890 77620-892 77620-894
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalonitrile ≥98%
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₅BN₂O₂
MDL Number: MFCD18730484
CAS Number: 2246614-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR