(8-Chloronaphthalen-1-yl)boronic acid ≥97%

Supplier: Ambeed

A1211300-250MG A1211300-10G A1211300-1G A1211300-5G
77571-264EA 92 USD
77571-264 77866-670 77866-671 77866-672
(8-Chloronaphthalen-1-yl)boronic acid ≥97%
(8-Chloronaphthalen-1-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₈BClO₂
MDL Number: MFCD18411302
CAS Number: 2305022-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR