2-Chloro-5-methoxybenzenesulfonyl chloride ≥98%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-5-methoxybenzene-1-sulphonyl chloride

A121105-100MG A121105-250MG A121105-5G
77685-065EA 35 USD
77685-065 77891-863 77891-864
2-Chloro-5-methoxybenzenesulfonyl chloride ≥98%
2-Chloro-5-methoxybenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆Cl₂O₃S
MDL Number: MFCD12026184
CAS Number: 201935-41-3
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR