(S,E)-3-(3,4-Dihydroxyphenyl)-2-(3-(3,4-dihydroxyphenyl)acrylamido)propanoic acid ≥97%

Supplier: Ambeed
Warning

A1210988-1MG A1210988-10MG A1210988-5MG A1210988-100MG A1210988-250MG A1210988-25MG A1210988-50MG
77596-330EA 30.75 USD
77596-330 77620-876 77620-878 77866-664 77866-665 77866-666 77866-667
(S,E)-3-(3,4-Dihydroxyphenyl)-2-(3-(3,4-dihydroxyphenyl)acrylamido)propanoic acid ≥97%
(S,E)-3-(3,4-Dihydroxyphenyl)-2-(3-(3,4-dihydroxyphenyl)acrylamido)propanoic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₇NO₇
MDL Number: MFCD18252514
CAS Number: 53755-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR