Benzothiazole-6-boronic acid, pinacol ester ≥98%

Supplier: Ambeed
Warning

A121051-1G A121051-250MG
77880-980EA 52 USD
77880-980 77880-981
Benzothiazole-6-boronic acid, pinacol ester ≥98%
Benzothiazole-6-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BNO₂S
MDL Number: MFCD12408395
CAS Number: 1002309-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR