2-Deoxy-2-fluoro-L-fucose ≥98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dideoxy-2-fluoro-L-galactose, (2S,3R,4R,5S)-2-Fluoro-3,4,5-trihydroxyhexanal

A1210299-1MG A1210299-10MG A1210299-5MG
77596-310EA 85.08 USD
77596-310 77620-870 77620-872
2-Deoxy-2-fluoro-L-fucose ≥98%
2-Deoxy-2-fluoro-L-fucose

Precaución: for Research Use Only

Formula: C₆H₁₁FO₄
MW: 166.148 g/mol
MDL Number: MFCD08457748
CAS Number: 70763-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR