BPMO ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(tert-Butoxycarbonyl)-2-methyl-3,4-dihydro-2H-pyrrole 1-oxide, 5-tert-Butoxycarbonyl-5-methyl-1-pyrroline-N-oxide, BMPO
A1210251-250MG
A1210251-1G
A1210251-50MG
77596-308EA
459.2
USD
77596-308
77620-866
77620-868
BPMO ≥97%
BMPO
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₃ MW: 199.3 g/mol |
MDL Number:
MFCD32196548 CAS Number: 387334-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...