5-Chloro-2-(9H-fluoren-9-yl)-4-(4-methoxyphenoxy)pyridazin-3(2H)-one ≥97%

Supplier: Ambeed

A1209830-10MG A1209830-1MG A1209830-5MG
77596-292EA 116.85 USD
77596-292 77596-294 77596-296
5-Chloro-2-(9H-fluoren-9-yl)-4-(4-methoxyphenoxy)pyridazin-3(2H)-one ≥97%
5-Chloro-2-(9H-fluoren-9-yl)-4-(4-methoxyphenoxy)pyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₂₄H₁₇ClN₂O₃
CAS Number: 1421365-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR