1-(4-Bromo-2-(1H-tetrazol-5-yl)phenyl)-3-(3-(trifluoromethyl)phenyl)urea 99+%

Supplier: Ambeed
Danger

Synonyms: NS3623

A1209642-10MG A1209642-1MG A1209642-5MG
77596-286EA 59.45 USD
77596-286 77596-288 77596-290
1-(4-Bromo-2-(1H-tetrazol-5-yl)phenyl)-3-(3-(trifluoromethyl)phenyl)urea 99+%
1-(4-Bromo-2-(1H-tetrazol-5-yl)phenyl)-3-(3-(trifluoromethyl)phenyl)urea

Precaución: for Research Use Only

Formula: C₁₅H₁₀BrF₃N₆O
MW: 427.1787 g/mol
MDL Number: MFCD22683830
CAS Number: 343630-41-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR