N-(3-Oxooctanoyl)-L-homoserine lactone ≥98%

Supplier: Ambeed
Warning

Synonyms: N-3-Oxo-octanoyl-L-homoserine lactone, (S)-4-Oxo-N-(2-oxotetrahydrofuran-3-yl)nonanamide, 3-oxo-C8-HSL

A1209560-10MG A1209560-5MG
77620-856EA 86.52 USD
77620-856 77620-858
N-(3-Oxooctanoyl)-L-homoserine lactone ≥98%
N-(3-Oxooctanoyl)-L-homoserine lactone

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₄
MW: 241.3 g/mol
MDL Number: MFCD08277025
CAS Number: 147795-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR