Z-Asp-2,6-dichlorobenzoyloxymethylketone ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-3-(((Benzyloxy)carbonyl)amino)-5-((2,6-dichlorobenzoyl)oxy)-4-oxopentanoic acid, Z-Asp-CH2-DCB

A1209363-10MG A1209363-1MG A1209363-5MG
77596-272EA 100.45 USD
77596-272 77596-274 77596-276
Z-Asp-2,6-dichlorobenzoyloxymethylketone ≥98%
Z-Asp-2,6-dichlorobenzoyloxymethylketone

Precaución: for Research Use Only

Formula: C₂₀H₁₇Cl₂NO₇
MW: 454.256 g/mol
MDL Number: MFCD00674885
CAS Number: 153088-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR