1-(4,6-Diamino-1,3,5-triazin-2-yl)guanidine ≥97%

Supplier: Ambeed

A1208547-100MG A1208547-50MG
77571-250EA 212 USD
77571-250 77596-266
1-(4,6-Diamino-1,3,5-triazin-2-yl)guanidine ≥97%
1-(4,6-Diamino-1,3,5-triazin-2-yl)guanidine

Precaución: for Research Use Only

Formula: C₄H₈N₈
MDL Number: MFCD08063773
CAS Number: 4405-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR