tert-Butyl 11-aminoundecanoate ≥97%

Supplier: Ambeed
Danger

A1208523-250MG A1208523-1G
77571-248EA 64.89 USD
77571-248 77592-354
tert-Butyl 11-aminoundecanoate ≥97%
tert-Butyl 11-aminoundecanoate

Precaución: for Research Use Only

Formula: C₁₅H₃₁NO₂
MW: 257.4121 g/mol
MDL Number: MFCD31729305
CAS Number: 220851-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR