1-Methyl-1H-indazole-7-carbaldehyde ≥98%

Supplier: Ambeed
Warning

A120811-100MG A120811-1G A120811-250MG
77871-529EA 50 USD
77871-529 77871-530 77871-531
1-Methyl-1H-indazole-7-carbaldehyde ≥98%
1-Methyl-1H-indazole-7-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MDL Number: MFCD11111019
CAS Number: 951030-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR